Merck
  • Investigation and characterization of liquid two-phase systems for the separation of crystal mixtures by interfacial partitioning.
Investigation and characterization of liquid two-phase systems for the separation of crystal mixtures by interfacial partitioning.
Biotechnology and bioengineering (2006-01-06)
Martijn A Hoeben, Pim van Hee, Rob G J M van der Lans, Gerard Kwant, Luuk A M van der Wielen
ABSTRACT
The interfacial partitioning behavior of ampicillin and phenylglycine crystals in different two-phase systems has been investigated. The two-phase systems employed are water/dodecane, water/1-butanol, and water/pentane/methanol. By means of partition experiments and microscopic imaging, it has been shown that the mechanism of separation strongly depends on the choice of the two-phase system. While water/dodecane features a mechanism of sheer competitive adsorption at the interface, separation in water/1-butanol is mainly due to partitioning into both liquid phases, leading to a higher degree of separation. Experiments with water/pentane/methanol have illustrated the large potential of three-component systems, as slight variations in the composition can have large effects on the separation.
Sigma-Aldrich
2-Phenylglycine, 95%
Sigma-Aldrich
L−(+)-α-Phenylglycine, 99%
Sigma-Aldrich
D−(−)-α-Phenylglycine, 99%