Merck
  • Simple chip-based interfaces for on-line monitoring of supramolecular interactions by nano-ESI MS.
Simple chip-based interfaces for on-line monitoring of supramolecular interactions by nano-ESI MS.
Lab on a chip (2005-09-22)
Monica Brivio, R Edwin Oosterbroek, Willem Verboom, Albert van den Berg, David N Reinhoudt
ABSTRACT
Two simple interfaces were designed and realized, enabling on-line coupling of microfluidics reactor chips to a nanoflow electrospray ionization (NESI) time-of-flight (TOF) mass spectrometer (MS). The interfaces are based on two different approaches: a monolithically integrated design, in which ionization is assisted by on-chip gas nebulization, and a modular approach implying the use of commercially available Picospray tips. Using reserpine as a reference compound in a 1ratio1 mixture of acetonitrile and water revealed that both interfaces provide a remarkably stable mass spectrometric signal (standard deviations lower than 8% and 1% for the monolithic and modular approaches, respectively). Glass microreactors, containing mixing zones, were fabricated and coupled to the modular interface with perfluoroelastomer Nanoport fluidics connectors, providing a tool to study chemical reactions on-line. Investigation of the mixing dynamics showed that complete on-chip reagents mixing is achieved within a few tens of milliseconds. Metal-ligand interactions of Zn-porphyrin with pyridine (2), 4-ethylpyridine (3), 4-phenylpyridine (4), N-methylimidazole (5), and N-butylimidazole (6) in acetonitrile as well as host-guest complexations of beta-cyclodextrin (7) with N-(1-adamantyl)acetamide (8) or 4-tert-butylacetanilide (9) in water were studied by mass spectrometry using the modular NESI-chip interface. From on-chip dilution-based mass spectrometric titrations of Zn-porphyrin 1 with pyridine (2) or 4-phenylpyridine (4) in acetonitrile Ka-values of 4.6 +/- 0.4 x 10(3) M(-1) and 6.5 +/- 1.2 x 10(3) M(-1), respectively, were calculated. The Ka-values are about four times larger than those obtained with UV/vis spectroscopy in solution, probably due to a higher ionization efficiency of complexed compared to uncomplexed Zn-porphyrin. For the complexation of N-(1-adamantyl)acetamide (8) with beta-cyclodextrin (7), a Ka-value of 3.6 +/- 0.3 x 10(4) M(-1) was obtained, which is in good agreement with that determined by microcalorimetry.
Sigma-Aldrich
1-Butylimidazole, 98%
Sigma-Aldrich
4-Ethylpyridine, 98%
Sigma-Aldrich
4-Phenylpyridine, 97%