Merck
  • Semiochemistry of the goldeneyed lacewing Chrysopa oculata: attraction of males to a male-produced pheromone.
Semiochemistry of the goldeneyed lacewing Chrysopa oculata: attraction of males to a male-produced pheromone.
Journal of chemical ecology (2004-12-14)
Qing-He Zhang, Kamlesh R Chauhan, Eric F Erbe, Ajay R Vellore, Jeffrey R Aldrich
ABSTRACT
Gas chromatographic-electroantennographic detection (G3C-EAD) experiments showed that antennae of males and females of the goldeneyed lacewing, Chrysopa oculata Say (Co. = Chrysopa), consistently responded to four compounds extracted from the abdominal cuticle of males:nonanal, nonanol, nonanoic acid, and (1R*,2S*,5R*,8R*)-iridodial. These compounds were not detected from abdominal cuticle of females. Thoracic extracts of both sexes contained antennal-stimulatory 1-tridecene and EAD-inactive skatole. Chrysopa oculata adults were most sensitive to (1R,2S,5R,8R)-iridodial standard at an EAD-response threshold between 0.1 and 1 pg, which was 10-100 times lower than thresholds for nonanal and nonanoic acid, and up to 10,000 times lower than thresholds for other compounds tested. A similar EAD response pattern was also found in another Chrysopa sp. (Co. quadripunctata Burmeister). In field-trapping experiments, (1R,2S,5R,8R)-iridodial was the only male-specific compound that attracted Co. oculata males. Males also were weakly attracted to (1R,4aS,7S,7aR)-nepetalactol (an aphid sex pheromone component), probably due to the 5% (1R,2S,5R,8R)-iridodial present in the synthetic sample as an impurity. A herbivore-induced plant volatile, methyl salicylate, increased attraction of males to (1R,2S,5R,8R)-iridodial, whereas 1-tridecene was antagonistic. No females were caught in the entire study. Scanning electron micrographs revealed numerous male-specific, elliptical epidermal glands on the 3rd-8th abdominal sternites of Co. oculata, which are likely the pheromone glands. Another lacewing species, Chrysoperla rufilabris (Burmeister) (Cl. = Chrysoperla), did not produce male-specific volatiles or possess the type of gland presumed to produce pheromone in Co. oculata males, but (Z)-4-tridecene was identified as a major antennal-stimulatory compound from thoracic extracts of both sexes of Cl. rufilabris. Thus, (1R,2S,5R,8R)-iridodial (or its enantiomer) is now identified as a male-produced male aggregation pheromone for Co. oculata, the first pheromone identified for lacewings.
Sigma-Aldrich
Nonanoic acid, 96%
Sigma-Aldrich
Nonanoic acid, ≥97%
Sigma-Aldrich
1-Tridecene, 96%
Sigma-Aldrich
Nonanoic acid, ≥96%, FG
Supelco
Nonanoic acid, analytical standard