- Field efficacy and baseline toxicities of pyriproxifen, acetamiprid, and diafenthiuron against Bemisia tabaci Gennadius (Homoptera: Aleyrodidae) in Burkina Faso (West Africa).
Field efficacy and baseline toxicities of pyriproxifen, acetamiprid, and diafenthiuron against Bemisia tabaci Gennadius (Homoptera: Aleyrodidae) in Burkina Faso (West Africa).
Bioassays were conducted in 2001 and 2002 to estimate toxicities and dose-response relationships of 24 Bemisica tabaci Gennadius populations to pyriproxifen, acemitaprid, and diafenthiuron. LC50s ranging from 0.014 to 0.096 mgL(-1), 0.60 to 1.3 mgL(-1), and 3.5 to 6.7 mgL(-1) were observed respectively for pyriproxifen, acemitaprid, and diafenthiuron. These LC50s much lower than the field doses recommended for each compound. A fast increase in rates of mortality within a narrow range of lethal concentrations was observed for each compound. indicating that all three compounds were highly effective at killing whiteflies. In a separate experiment, pyriproxifen, acemitaprid, and diafenthiuron were tested in 2001 and 2002 to compare their effectiveness and assess their impact on parasitism in the field. In both years all three compounds significantly prevented B. tabaci populations from reaching economic injury levels in cotton and minimized adverse effects on parasitism. Our results provide for the first time baseline toxicological, field efficacy, and effect on parasitism data for pyriproxifen, acemitaprid, and diafenthiuron against B. tabaci in West Africa. These compounds should be included in a resistance management program of the cotton pest complex and their use should be restricted to prevent the building of resistance in B. tabaci populations.