Merck
  • Amino acid substitutions in norovirus VP1 dictate host dissemination via variations in cellular attachment.
Amino acid substitutions in norovirus VP1 dictate host dissemination via variations in cellular attachment.
Journal of virology (2023-11-30)
Jake T Mills, Susanna C Minogue, Joseph S Snowden, Wynter K C Arden, David J Rowlands, Nicola J Stonehouse, Christiane E Wobus, Morgan R Herod
ABSTRACT
All viruses initiate infection by utilizing receptors to attach to target host cells. These virus-receptor interactions can therefore dictate viral replication and pathogenesis. Understanding the nature of virus-receptor interactions could also be important for the development of novel therapies. Noroviruses are non-enveloped icosahedral viruses of medical importance. They are a common cause of acute gastroenteritis with no approved vaccine or therapy and are a tractable model for studying fundamental virus biology. In this study, we utilized the murine norovirus model system to show that variation in a single amino acid of the major capsid protein alone can affect viral infectivity through improved attachment to suspension cells. Modulating plasma membrane mobility reduced infectivity, suggesting an importance of membrane mobility for receptor recruitment and/or receptor conformation. Furthermore, different substitutions at this site altered viral tissue distribution in a murine model, illustrating how in-host capsid evolution could influence viral infectivity and/or immune evasion.
Sigma-Aldrich
Anti-Norovirus (MNV-1) Antibody, clone 5C4.10, clone 5C4.10, from mouse
Pricing and availability is not currently available.