Merck
  • SorCS1 inhibits amyloid-β binding to neurexin and rescues amyloid-β-induced synaptic pathology.
SorCS1 inhibits amyloid-β binding to neurexin and rescues amyloid-β-induced synaptic pathology.
Life science alliance (2023-01-26)
Alfred Kihoon Lee, Nayoung Yi, Husam Khaled, Benjamin Feller, Hideto Takahashi
ABSTRACT
Amyloid-β oligomers (AβOs), toxic peptide aggregates found in Alzheimer's disease, cause synapse pathology. AβOs interact with neurexins (NRXs), key synaptic organizers, and this interaction dampens normal trafficking and function of NRXs. Axonal trafficking of NRX is in part regulated by its interaction with SorCS1, a protein sorting receptor, but the impact of SorCS1 regulation of NRXs in Aβ pathology was previously unstudied. Here, we show competition between the SorCS1 ectodomain and AβOs for β-NRX binding and rescue effects of the SorCS1b isoform on AβO-induced synaptic pathology. Like AβOs, the SorCS1 ectodomain binds to NRX1β through the histidine-rich domain of NRX1β, and the SorCS1 ectodomain and AβOs compete for NRX1β binding. In cultured hippocampal neurons, SorCS1b colocalizes with NRX1β on the axon surface, and axonal expression of SorCS1b rescues AβO-induced impairment of NRX-mediated presynaptic organization and presynaptic vesicle recycling and AβO-induced structural defects in excitatory synapses. Thus, our data suggest a role for SorCS1 in the rescue of AβO-induced NRX dysfunction and synaptic pathology, providing the basis for a novel potential therapeutic strategy for Alzheimer's disease.
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Protein G Sepharose 4 Fast Flow, Cytiva 17-0618-01, pack of 5 mL