Merck
  • A small molecule inhibitor of the UBE2F-CRL5 axis induces apoptosis and radiosensitization in lung cancer.
A small molecule inhibitor of the UBE2F-CRL5 axis induces apoptosis and radiosensitization in lung cancer.
Signal transduction and targeted therapy (2022-10-18)
Tiantian Xu, Qisheng Ma, Yanan Li, Qing Yu, Peichen Pan, Yawen Zheng, Zhijian Li, Xiufang Xiong, Tingjun Hou, Bin Yu, Hongmin Liu, Yi Sun
ABSTRACT
Protein neddylation is catalyzed by a neddylation activating enzyme (NAE, E1), an E2 conjugating enzyme, and an E3 ligase. In various types of human cancers, the neddylation pathway is abnormally activated. Our previous study validated that the neddylation E2 UBE2F is a promising therapeutic target in lung cancer. Although the NAE inhibitor MLN4924/pevonedistat is currently under clinical investigation as an anti-cancer agent, there are no small molecules available that selectively target UBE2F. Here, we report, for the first time, the discovery, via structure-based virtual screen and chemical optimization, of such a small molecule, designated as HA-9104. HA-9104 binds to UBE2F, reduces its protein levels, and consequently inhibits cullin-5 neddylation. Blockage of cullin-5 neddylation inactivates cullin-RING ligase-5 (CRL5) activity, leading to accumulation of the CRL5 substrate, NOXA, to induce apoptosis. Moreover, HA-9104 appears to form the DNA adduct via its 7-azaindole group to induce DNA damage and G2/M arrest. Biologically, HA-9104 effectively suppresses the growth and survival of lung cancer cells and confers radiosensitization in both in vitro cell culture and in vivo xenograft tumor models. In summary, we discovered a small molecule, designated HA-9104, that targets the UBE2F-CRL5 axis with anti-cancer activity alone or in combination with radiation.
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-CUL5 antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Anti-Noxa Mouse mAb (114C307), liquid, clone 114C307, Calbiochem®
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone AA13, purified from hybridoma cell culture