Merck
  • Dopamine signaling impairs ROS modulation by mitochondrial hexokinase in human neural progenitor cells.
Dopamine signaling impairs ROS modulation by mitochondrial hexokinase in human neural progenitor cells.
Bioscience reports (2021-11-26)
Gabriela Assis-de-Lemos, Jamila Monteiro, Viviane M Oliveira-Valença, Guilherme A Melo, Ricardo A de Melo Reis, Stevens K Rehen, Mariana S Silveira, Antonio Galina
ABSTRACT
Dopamine signaling has numerous roles during brain development. In addition, alterations in dopamine signaling may be also involved in the pathophysiology of psychiatric disorders. Neurodevelopment is modulated in multiple steps by reactive oxygen species (ROS), byproducts of oxidative metabolism that are signaling factors involved in proliferation, differentiation, and migration. Hexokinase (HK), when associated with the mitochondria (mt-HK), is a potent modulator of the generation of mitochondrial ROS in the brain. In the present study, we investigated whether dopamine could affect both the activity and redox function of mt-HK in human neural progenitor cells (NPCs). We found that dopamine signaling via D1R decreases mt-HK activity and impairs ROS modulation, which is followed by an expressive release of H2O2 and impairment in calcium handling by the mitochondria. Nevertheless, mitochondrial respiration is not affected, suggesting specificity for dopamine on mt-HK function. In neural stem cells (NSCs) derived from induced-pluripotent stem cells (iPSCs) of schizophrenia patients, mt-HK is unable to decrease mitochondrial ROS, in contrast with NSCs derived from healthy individuals. Our data point to mitochondrial hexokinase as a novel target of dopaminergic signaling, as well as a redox modulator in human neural progenitor cells, which may be relevant to the pathophysiology of neurodevelopmental disorders such as schizophrenia.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
Anti-Dopamine D₁ Receptor Rabbit pAb, lyophilized, Calbiochem®