Merck
  • Single-cell transcriptomic profiling provides insights into the toxic effects of Zearalenone exposure on primordial follicle assembly.
Single-cell transcriptomic profiling provides insights into the toxic effects of Zearalenone exposure on primordial follicle assembly.
Theranostics (2021-04-17)
Yu Tian, Ming-Yu Zhang, Ai-Hong Zhao, Li Kong, Jun-Jie Wang, Wei Shen, Lan Li
ABSTRACT
Rationale: Zearalenone (ZEN), a pollutant in our daily diet, seriously threatens the reproductive health of humans and animals. The primordial follicle (PF) assembly in the mouse occurs during the perinatal period, which determines the whole ovarian reserve in reproductive lifespan. In the current investigation, we administered ZEN orally to perinatal mice from 16.5 days post coitum (dpc) to postnatal day 3 (PD3), and single-cell RNA sequencing (scRNA-seq) was performed on PD0 and PD3 ovarian tissues in the offspring to check ZEN toxic to primordial follicle formation at the single cell level. Methods: Ovarian tissues (in vivo) were examined by single cell RNA sequencing analysis, Immunostaining, and Western blotting. Ovarian tissues (in vitro) were examined by qRT-PCR, Immunostaining, and Western blotting. Results: We found that ZEN exposure altered the developmental trajectory of both germ cells and granulosa cells. Furthermore, after establishing the cell-cell communication network between germ cells and granulosa cells, we found that this was perturbed by ZEN exposure, especially during the Hippo signaling pathway. Conclusions: This study showed that ZEN affected the status of germ cells and granulosa cells through the Hippo signaling pathway and blocked the assembly of PFs. This research contributes to our deeper understanding of the mechanisms of toxicity in different cell types and the disruption of normal intercellular signaling by ZEN exposure.
Sigma-Aldrich
Zearalenone, fungal mycotoxin
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Bovine Serum Albumin, chromatographically purified, New Zealand origin, low endotoxin, suitable for cell culture, pH 7, ≥98%