Merck
Recent advances in microchip enantioseparation and analysis.
Electrophoresis (2020-09-20)
Nan Lu, Jörg P Kutter
ABSTRACT
This review summarizes recent developments (over the past decade) in the field of microfluidics-based solutions for enantiomeric separation and detection. The progress in various formats of microchip electrodriven separations, such as MCE, microchip electrochromatography, and multidimensional separation techniques, is discussed. Innovations covering chiral stationary phases, surface coatings, and modification strategies to improve resolution, as well as integration with detection systems, are reported. Finally, combinations with other microfluidic functional units are also presented and highlighted.
Sigma-Aldrich
3,4-Dihydroxy-D-phenylalanine, powder, ≥95%