Merck
  • Repurposing Fenamic Acid Drugs To Combat Multidrug-Resistant Neisseria gonorrhoeae.
Repurposing Fenamic Acid Drugs To Combat Multidrug-Resistant Neisseria gonorrhoeae.
Antimicrobial agents and chemotherapy (2020-05-13)
Young Jin Seong, Marwa Alhashimi, Abdelrahman Mayhoub, Haroon Mohammad, Mohamed N Seleem
ABSTRACT
The rise of extensively drug-resistant and multidrug-resistant strains of Neisseria gonorrhoeae has occurred in parallel with the increasing demand for new drugs. However, the current methods of drug discovery are burdened with rigorous assessments and require more time than can be spared until gonococcal infections become difficult to control. To address this urgency, we utilized a drug-repurposing strategy and identified three clinically approved anthranilic acid drugs (tolfenamic acid, flufenamic acid, and meclofenamic acid) with potent antigonococcal activity, inhibiting 50% of the strains (MIC50) from 4 to 16 μg/ml. Furthermore, tolfenamic acid showed indifferent activity with antibiotics of choice for gonococcal infections, azithromycin and ceftriaxone, in checkerboard assays with a fractional inhibitory concentration index ranging from 0.75 to 1.5. Fenamic acids reduced a high inoculum of N. gonorrhoeae below the limit of detection within 12 h and exhibited a low frequency of resistance. Interestingly, the fenamic acids did not inhibit the growth of commensal Lactobacillus spp. that comprise the healthy female genital microbiota. Fenamic acids were also superior to ceftriaxone in reducing the burden of intracellular N. gonorrhoeae within infected endocervical cells by 99%. Furthermore, all three fenamic acids significantly reduced the expression of proinflammatory cytokines by infected endocervical cells. Finally, fenamic acids and other structurally related anthranilic acid derivatives were evaluated to ascertain a more in-depth structure-activity relationship (SAR) that revealed N-phenylanthranilic acid as a novel antigonorrheal scaffold. This SAR study will pave the road to repositioning more potent fenamic acids analogues against N. gonorrhoeae.
Sigma-Aldrich
N-(2-Amino-4-chlorophenyl)anthranilic acid, 97%
Sigma-Aldrich
Hematin porcine
Sigma-Aldrich
3-amino-2-phenylamino-benzoic acid, AldrichCPR
Supelco
Flufenamic acid, analytical standard, for drug analysis