Merck
  • An androgen-independent mechanism underlying the androgenic effects of 3-methylcholanthrene, a potent aryl hydrocarbon receptor agonist.
An androgen-independent mechanism underlying the androgenic effects of 3-methylcholanthrene, a potent aryl hydrocarbon receptor agonist.
Toxicology research (2020-07-17)
Noriko Sanada, Yuka Gotoh-Kinoshita, Naoya Yamashita, Ryoichi Kizu
ABSTRACT
Aryl hydrocarbon receptor (AhR) and androgen receptor (AR) are ligand-activated transcription factors with profound cross-talk between their signal transduction pathways. Previous studies have shown that AhR agonists activate the transcription of AR-regulated genes in an androgen-independent manner; however, the underlying mechanism remains unclear. To decipher this mechanism, we evaluated the effects of 3-methylcholanthrene (3MC), a potent AhR agonist, on the transcription of AR-regulated genes in three AR-expressing cell lines. 3MC induced the expression of not only three representative AR-regulated chromosomal genes but also the exogenous AR-responsive luciferase reporter gene. No significant difference in the 3MC-induced luciferase activity was detected in the presence of SKF-525A, a non-specific inhibitor of CYP enzymes. The androgenic effects of 3MC were diminished by AhR and AR knockdown. Following 3MC treatment, the amount of nuclear AhR and AR increased synchronously. Co-immunoprecipitation revealed that AhR and AR formed a complex in the nucleus of cells treated with 3MC. AR was recruited to the proximal promoter and distal enhancer regions of the PSA gene upon the addition of 3MC. We propose that AhR activated by 3MC forms a complex with unliganded AR which translocates from the cytoplasm to the nucleus. Nuclear AR now binds the transcriptional regulatory region of AR-regulated genes and activates the transcription.
Sigma-Aldrich
Anti-Androgen Receptor Antibody, from rabbit, purified by affinity chromatography
Supelco
Proadifen hydrochloride, analytical standard, ≥95%
Sigma-Aldrich
Anti-RPLP0 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Histone H2B Antibody, Upstate®, from rabbit