- Homodimerization and intermolecular tyrosine phosphorylation of the Tyk-2 tyrosine kinase.
Homodimerization and intermolecular tyrosine phosphorylation of the Tyk-2 tyrosine kinase.
FEBS letters (1995-11-06)
P Domanski, H Yan, M M Witte, J Krolewski, O R Colamonici
PMID7589562
ABSTRACT
The Jak kinases and Stat transcription factors play a major role in signaling of various cytokines including IFN alpha. In this report we show a ligand-independent interaction between Tyk-2 and Jak-1 kinases. We also demonstrate that the Tyk-2 kinase forms a homodimer that has the ability to undergo intermolecular tyrosine phosphorylation. The formation of the Tyk-2 homodimer is independent of both tyrosine phosphorylation and the presence of the tyrosine kinase domain.
Sigma-Aldrich
Tyk2 active human, recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE)