Merck
  • Effect of HIV-1 TAT Peptide Fusion on 5' mRNA Cap Analogs Cell Membrane Permeability and Translation Inhibition.
Effect of HIV-1 TAT Peptide Fusion on 5' mRNA Cap Analogs Cell Membrane Permeability and Translation Inhibition.
Bioconjugate chemistry (2020-04-02)
Karolina Piecyk, Paulina Pietrow, Thomas Arnold, Remigiusz Worch, Nadejda L Korneeva, Marzena Jankowska-Anyszka
ABSTRACT
The development of targeted anticancer drugs has been one of the most challenging goals of current research. Eukaryotic translation initiation factor 4E (eIF4E) is an oncogene that stimulates mRNA translation via binding to the 5' endcap structure. It is well documented that eIF4E is overexpressed in many cancers including breast, prostate, head and neck, and stomach malignancies and leads to oncogenic transformation and metastasis. One approach to block eIF4E function in cancer cells is based on the disruption of the interaction between eIF4E and the 5' mRNA cap structure using cap analog inhibitors. Since analogs are cell-impermeable due to their anionic nature, we used a cell penetrating peptide (CPP) for delivery of model cap analogs into cancer cells. The human immunodeficiency virus I (HIV-1) transactivator of transcription derived peptide (TAT) was conjugated with the analogs m7GMP and m7GpppG using click chemistry methodology. We observed that both conjugates (m7GMP-TAT and m7GpppG-TAT), contrary to TAT alone, did not translocate through the artificial phospholipid membrane of giant unilamellar vesicles. This suggests that passive transport is not the mechanism by which translocation of cap analogs occurs. In contrast, synthesized fluorescently labeled m7GpppG-TAT translocated into the human breast adenocarcinoma cancer cell line MCF-7. Furthermore, we demonstrated that m7GMP-TAT and m7GpppG-TAT inhibited cap-dependent translation up to 30% both in vivo and in vitro while simultaneously not affecting cell growth and viability. These results demonstrate the usefulness of cell penetration peptides as carriers for the internalization of cap analogs.
Product Description
Avanti
16:0-06:0 NBD PC, Avanti Research - A Croda Brand 810130C
Avanti
16:0-06:0 NBD PC, Avanti Research - A Croda Brand 810130P, powder
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand