Merck
  • Human ESC-Derived Chimeric Mouse Models of Huntington's Disease Reveal Cell-Intrinsic Defects in Glial Progenitor Cell Differentiation.
Human ESC-Derived Chimeric Mouse Models of Huntington's Disease Reveal Cell-Intrinsic Defects in Glial Progenitor Cell Differentiation.
Cell stem cell (2018-12-18)
Mikhail Osipovitch, Andrea Asenjo Martinez, John N Mariani, Adam Cornwell, Simrat Dhaliwal, Lisa Zou, Devin Chandler-Militello, Su Wang, Xiaojie Li, Sarah-Jehanne Benraiss, Robert Agate, Andrea Lampp, Abdellatif Benraiss, Martha S Windrem, Steven A Goldman
ABSTRACT
Huntington's disease (HD) is characterized by hypomyelination and neuronal loss. To assess the basis for myelin loss in HD, we generated bipotential glial progenitor cells (GPCs) from human embryonic stem cells (hESCs) derived from mutant Huntingtin (mHTT) embryos or normal controls and performed RNA sequencing (RNA-seq) to assess mHTT-dependent changes in gene expression. In human GPCs (hGPCs) derived from 3 mHTT hESC lines, transcription factors associated with glial differentiation and myelin synthesis were sharply downregulated relative to normal hESC GPCs; NKX2.2, OLIG2, SOX10, MYRF, and their downstream targets were all suppressed. Accordingly, when mHTT hGPCs were transplanted into hypomyelinated shiverer mice, the resultant glial chimeras were hypomyelinated; this defect could be rescued by forced expression of SOX10 and MYRF by mHTT hGPCs. The mHTT hGPCs also manifested impaired astrocytic differentiation and developed abnormal fiber architecture. White matter involution in HD is thus a product of the cell-autonomous, mHTT-dependent suppression of glial differentiation.
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder
Sigma-Aldrich
Anti-Nuclei Antibody, clone 235-1, clone 235-1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Nuclei Antibody, clone 235-1, Alexa Fluor 488 conjugate, clone 235-1, from mouse, ALEXA FLUOR 488