Merck
  • The p85beta regulatory subunit of PI3K serves as a substrate for PTEN protein phosphatase activity during insulin mediated signaling.
The p85beta regulatory subunit of PI3K serves as a substrate for PTEN protein phosphatase activity during insulin mediated signaling.
Biochemical and biophysical research communications (2010-06-03)
Jiman He, Suzanne de la Monte, Jack R Wands
ABSTRACT
Phosphatase and tensin homolog deleted on chromosome 10 (PTEN) is a tumor suppressor gene that is frequently mutated in brain, uterine, and prostate cancers. The protein phosphatase activity is poorly defined. We demonstrate that insulin stimulates phosphorylation of tyrosine and threonine/proline residues on the p85 regulatory subunit of PI3K in Huh-7, and HEK 293 cells. The specificity of PTEN binding and dephosphorylation of PI3K appears to reside on the p85beta subunit. Therefore, the PTEN phosphatase is active against the PI3K p85beta subunit and dephosphorylates a protein involved in insulin signaling where known downstream consequences are increased cell migration, motility, and invasion.
Sigma-Aldrich
Anti-HA Tag Antibody, Upstate®, from mouse
Sigma-Aldrich
Anti-Phosphothreonine Antibody, Chemicon®, from rabbit