Merck
  • PDGF-mediated mesenchymal transformation renders endothelial resistance to anti-VEGF treatment in glioblastoma.
PDGF-mediated mesenchymal transformation renders endothelial resistance to anti-VEGF treatment in glioblastoma.
Nature communications (2018-08-29)
Tianrun Liu, Wenjuan Ma, Haineng Xu, Menggui Huang, Duo Zhang, Zhenqiang He, Lin Zhang, Steven Brem, Donald M O'Rourke, Yanqing Gong, Yonggao Mou, Zhenfeng Zhang, Yi Fan
ABSTRACT
Angiogenesis is a hallmark of cancer. However, most malignant solid tumors exhibit robust resistance to current anti-angiogenic therapies that primarily target VEGF pathways. Here we report that endothelial-mesenchymal transformation induces glioblastoma (GBM) resistance to anti-angiogenic therapy by downregulating VEGFR-2 expression in tumor-associated endothelial cells (ECs). We show that VEGFR-2 expression is markedly reduced in human and mouse GBM ECs. Transcriptome analysis verifies reduced VEGFR-2 expression in ECs under GBM conditions and shows increased mesenchymal gene expression in these cells. Furthermore, we identify a PDGF/NF-κB/Snail axis that induces mesenchymal transformation and reduces VEGFR-2 expression in ECs. Finally, dual inhibition of VEGFR and PDGFR eliminates tumor-associated ECs and improves animal survival in GBM-bearing mice. Notably, EC-specific knockout of PDGFR-β sensitizes tumors to VEGF-neutralizing treatment. These findings reveal an endothelial plasticity-mediated mechanism that controls anti-angiogenic therapy resistance, and suggest that vascular de-transformation may offer promising opportunities for anti-vascular therapy in cancer.
Sigma-Aldrich
Anti-PDGF-AA Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-NG2 Chondroitin Sulfate Proteoglycan Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-PDGF-BB Antibody, from rabbit, purified by affinity chromatography