Merck
  • Class IIa HDACs regulate learning and memory through dynamic experience-dependent repression of transcription.
Class IIa HDACs regulate learning and memory through dynamic experience-dependent repression of transcription.
Nature communications (2019-08-04)
Yongchuan Zhu, Min Huang, Eric Bushong, Sebastien Phan, Marco Uytiepo, Elizabeth Beutter, Daniel Boemer, Kristin Tsui, Mark Ellisman, Anton Maximov
ABSTRACT
The formation of new memories requires transcription. However, the mechanisms that limit signaling of relevant gene programs in space and time for precision of information coding remain poorly understood. We found that, during learning, the cellular patterns of expression of early response genes (ERGs) are regulated by class IIa HDACs 4 and 5, transcriptional repressors that transiently enter neuronal nuclei from cytoplasm after sensory input. Mice lacking these repressors in the forebrain have abnormally broad experience-dependent expression of ERGs, altered synaptic architecture and function, elevated anxiety, and severely impaired memory. By acutely manipulating the nuclear activity of class IIa HDACs in behaving animals using a chemical-genetic technique, we further demonstrate that rapid induction of transcriptional programs is critical for memory acquisition but these programs may become dispensable when a stable memory is formed. These results provide new insights into the molecular basis of memory storage.
Sigma-Aldrich
Anti-Histone Deacetylase 4 (HDAC4) (ML-19) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-β-Tubulin III antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Histone Deacetylase 5 (HDAC5) antibody, Mouse monoclonal, clone HDAC5-35, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Prox 1 Antibody, serum, Chemicon®