Merck
  • Helicobacter pylori VacA induces autophagic cell death in gastric epithelial cells via the endoplasmic reticulum stress pathway.
Helicobacter pylori VacA induces autophagic cell death in gastric epithelial cells via the endoplasmic reticulum stress pathway.
Cell death & disease (2017-12-15)
Pan Zhu, Jun Xue, Zhu-Jun Zhang, Yin-Ping Jia, Ya-Nan Tong, Dan Han, Qian Li, Yang Xiang, Xu-Hu Mao, Bin Tang
ABSTRACT
The Helicobacter pylori vacuolating cytotoxin (VacA) can promote progressive vacuolation and gastric injury and may be associated with human gastric cancer. Increasing evidence indicates that autophagy is involved in the cell death induced by VacA, but the specific mechanisms need to be further elucidated. We show here that VacA could induce autophagy and increase cell death in human gastric cancer cell lines. Further investigations revealed that inhibition of autophagy could decrease the VacA-induced cell death in AGS cells. Furthermore, numerous dilated endoplasmic reticula (ER) were observed, and the phosphorylation of a subunit of eukaryotic translation initiation factor 2 subunit 1 also increased in the VacA-treated AGS cells, while repression of ER stress could reduce autophagy and cell death through knockdown of activating transcription factor 4 and DNA-damage-inducible transcript 3. In addition, the expression of pseudokinase tribbles homolog 3 (TRIB3) upon ER stress was triggered by VacA, and knockdown of TRIB3 could also decrease VacA-induced cell death. Finally, inhibition of autophagy could decrease VacA s1m1 -induced cell death and apoptosis, and apoptosis inhibitor Z-VAD had no significant effect on autophagy induced by VacA s1m1 . Thus, these results suggested that VacA causes autophagic cell death via ER stress in gastric epithelial cells.
Sigma-Aldrich
Monoclonal Anti-TRIB3 antibody produced in mouse, clone 1H2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Phosphate buffered saline, 10× concentrate, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Z-VAD-FMK, solid