
Valaciclovir impurity G

Synonym(s):
4-(Dimethylamino)pyridine, N,N-Dimethylpyridin-4-amine, DMAP
Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing