
Tridodecylmethylammonium chloride

Synonym(s):
Methyltridodecylammonium chloride
Linear Formula:
[CH3(CH2)11]3N(Cl)CH3
CAS Number:
Molecular Weight:
572.47
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing