
Tiotropium bromide monohydrate

Empirical Formula (Hill Notation):
C19H22BrNO4S2 · H2O
CAS Number:
Molecular Weight:
490.43
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing