
SUPELCOGEL™ 8Ca (9 µm) HPLC Columns

Synonym(s):
SUPELCOGEL™ 6% Cross Linked HPLC Columns, SUPELCOGEL™ 8% Crosslinked HPLC Column
Products
Product Category
Brand
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Column Material
Package Size
Particle Size (µm)
Technique
Showing 1-3 of 3