
Sulfasalazine

Synonym(s):
2-Hydroxy-5-{{4-[(2-pyridinylamino)sulfonyl]phenyl}azo}benzoic acid, 5-[4-(2-Pyridylsulfamoyl)phenylazo]salicylic acid, Salicylazosulfapyridine, Sulfasalazine
Empirical Formula (Hill Notation):
C18H14N4O5S
CAS Number:
Molecular Weight:
398.39
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4