![Saflufenacil certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/208/972/9006566a-4f44-4334-aa9e-188c4af17ee0/640/9006566a-4f44-4334-aa9e-188c4af17ee0.png)
Saflufenacil
![Saflufenacil certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/208/972/9006566a-4f44-4334-aa9e-188c4af17ee0/640/9006566a-4f44-4334-aa9e-188c4af17ee0.png)
Synonym(s):
2-Chloro-5-[3,6-dihydro-3-methyl-2,6-dioxo-4-(trifluoromethyl)-1(2H)-pyrimidinyl]-4-fluoro-N-{[methyl(1-methylethyl)amino]sulfonyl}benzamide, N′-{2-Chloro-4-fluoro-5-[1,2,3,6-tetrahydro-3-methyl-2,6-dioxo-4-(trifluoromethyl)pyrimidin-1-yl]benzoyl}-N-isopropyl-N-methylsulfamide
Empirical Formula (Hill Notation):
C17H17ClF4N4O5S
CAS Number:
Molecular Weight:
500.85
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2