
Rebaudioside E

Synonym(s):
(-)-Rebaudioside E, Kaur-16-en-18-oic acid, 13-[(2-O-β-D-glucopyranosyl-β-D-glucopyranosyl)oxy]-, 2-O-β-D-glucopyranosyl-β-D-glucopyranosyl ester, (4α)-
Empirical Formula (Hill Notation):
C44H70O23
CAS Number:
Molecular Weight:
967.01
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing