
Piperazine hydrate

Synonym(s):
Piperazine hexahydrate
Empirical Formula (Hill Notation):
C4H10N2 · 6H2O
CAS Number:
Molecular Weight:
194.23
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing