
Perphenazine sulfoxide

Synonym(s):
4-[3-(2-Chlorophenothiazin-10-yl)propyl]-1-piperazineethanol sulfoxide1-Piperazineethanol sulfoxide
Empirical Formula (Hill Notation):
C21H26ClN3O2S
CAS Number:
Molecular Weight:
419.97
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2