
Palonosetron Related Compound E

Synonym(s):
2-[(3S)-1-Azabicyclo[2.2.2]oct-3-yl]-2,4,5,6-tetrahydro-1H-benz[de]isoquinolin-1-one monohydrochloride, 2-[(3S)-Quinuclidin-3-yl]-2,4,5,6-tetrahydro-1H-benzo[de]isoquinolin-1-one hydrochloride
Empirical Formula (Hill Notation):
C19H22N2O · HCl
CAS Number:
Molecular Weight:
330.85
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing