
Nigericin sodium salt

Synonym(s):
Antibiotic K178, Antibiotic X464, Azalomycin M, Helexin C, Polyetherin A, 3B2-6379, HE331800, sodium;2-[(3S,6R)-6-[[(5R,6R,7R,9R)-2-[5-[(3S,5R)-5-[(2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3-methyloxolan-2-yl]-5-methyloxolan-2-yl]-7-methoxy-2,4,6-trimethyl-1,10-dioxaspiro[4.5]decan-9-yl]methyl]-3-methyloxan-2-yl]propanoate
Empirical Formula (Hill Notation):
C40H67NaO11
CAS Number:
Molecular Weight:
746.94
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Showing 1-2 of 2
Compare
Product Number
Description
Pricing