Skip to Content
Products
Cart
0
CX
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
N
-Ethylmaleimide
Synonym(s):
NEM
Empirical Formula (Hill Notation):
C
6
H
7
NO
2
CAS Number:
128-53-0
Molecular Weight:
125.13
EC Number:
204-892-4
Products
Brand
Sigma-Aldrich (4)
Biological Source
Boiling Point (°C)
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Special Grade
Purum (1)
Purum P.A. (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
04260
N
-Ethylmaleimide,
purum p.a., ≥99.0% (HPLC)
Expand
View Pricing
04259
N
-Ethylmaleimide,
BioUltra, ≥99.0% (HPLC)
Expand
View Pricing
E3876
N
-Ethylmaleimide,
crystalline, ≥98% (HPLC)
Expand
View Pricing
E1271
N
-Ethylmaleimide,
BioXtra, ≥98% (HPLC)
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Learn More