
Naringin

Synonym(s):
4′,5,7-Trihydroxyflavanone 7-rhamnoglucoside, Naringenin 7-neohesperidoside, Naringenine-7-rhamnosidoglucoside, Naringoside
Empirical Formula (Hill Notation):
C27H32O14
CAS Number:
Molecular Weight:
580.53
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Tissue
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5