
Mafenide acetate

Synonym(s):
4-Aminomethylbenzenesulfonamide acetate salt, 4-Homosulfanilamide, Mafenide, Marfanil
Empirical Formula (Hill Notation):
C7H10N2O2S · C2H4O2
CAS Number:
Molecular Weight:
246.28
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing