
L-(−)-Norepinephrine (+)-bitartrate salt monohydrate

Synonym(s):
L-(−)-Noradrenaline (+)-bitartrate salt monohydrate, L-(−)-Norepinephrine-(+)-bitartrate salt monohydrate
Empirical Formula (Hill Notation):
C8H11NO3 · C4H6O6 · H2O
CAS Number:
Molecular Weight:
337.28
Products
Product Category
Brand
Color
Feature
Formula Weight
Physical Form
Purity
Agency
Research Area
Quality Segment
Application
Optical Isomer
Showing 1-3 of 3
Compare
Product Number
Description
Pricing