Skip to Content
Merck
Linoleic acid

Linoleic acid

Linoleic acid
Synonym(s):
cis-9,cis-12-Octadecadienoic acid, Linoleic acid
Linear Formula:
CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H
CAS Number:
Molecular Weight:
280.45
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Physical Form
Purity
Agency
Research Area
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Special Grade
Technique