
L-(+)-Lactic acid

Synonym(s):
(S)-2-Hydroxypropionic acid, Sarcolactic acid
Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
Products
Product Category
Brand
Feature
Formula Weight
Physical Form
Product Line
Purity
Activity Spectrum
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Mode of Action
Optical Isomer
Package Size
API Family
Special Grade
Showing 1-5 of 5