
Kresoxim-methyl

Synonym(s):
Methyl (E)-α-(methoxyimino)-2-(2-methylphenoxymethyl)phenylacetate, Kresoxim-methyl
Empirical Formula (Hill Notation):
C18H19NO4
CAS Number:
Molecular Weight:
313.35
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2