
HEPES hemisodium salt

Synonym(s):
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid hemisodium salt, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) hemisodium salt
Empirical Formula (Hill Notation):
C8H17.5N2O4S· 0.5Na
CAS Number:
Molecular Weight:
249.30
Products
Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
pK Value
Purity
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing