
Harmane

Synonym(s):
1-Methyl-9H-pyrido[3,4-b]indole, 2-Methyl-β-carboline, Aribine
Empirical Formula (Hill Notation):
C12H10N2
CAS Number:
Molecular Weight:
182.22
Products
Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing