
Europium

Synonym(s):
Europium
Empirical Formula (Hill Notation):
Eu
CAS Number:
Molecular Weight:
151.96
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (mm)
Element
Showing 1-2 of 2
Compare
Product Number
Description
Pricing