Skip to Content
CX
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Estragole
Synonym(s):
4-Allylanisole,
p
-Allylphenyl methyl ether,
p
-Methoxyallylbenzene, Chavicol methyl ether, Estragole, Isoanethole, Methyl chavicol
Linear Formula:
H
2
C=CHCH
2
C
6
H
4
OCH
3
CAS Number:
140-67-0
Molecular Weight:
148.20
EC Number:
205-427-8
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
analytical standard (2)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
PHL82205
Estragole,
phyproof
®
Reference Substance
Expand
View Pricing
34098
Estragole,
analytical standard
Expand
View Pricing