
DMT-dI Phosphoramidite

Synonym(s):
DMT-dI amidite
Empirical Formula (Hill Notation):
C40H47N6O7P
CAS Number:
Molecular Weight:
754.81
Products
Product Category
Brand
Formula Weight
Product Line
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing