Dinoprost tromethamine
Synonym(s):
Prostaglandin F2α tris salt, (5Z,9α,11α,13E,15S)-9,11,15-Trihydroxyprosta-5,13-dienoic acid tris salt, PGF2α−Tris
Empirical Formula (Hill Notation):
C20H34O5 · C4H11NO3
CAS Number:
Molecular Weight:
475.62
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing