Skip to Content
Merck
Delphinidin chloride phyproof® Reference Substance

Delphinidin chloride

Delphinidin chloride phyproof® Reference Substance
Synonym(s):
3,3′,4′,5,5′,7-Hexahydroxyflavylium chloride, 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-1-benzopyrylium chloride
Empirical Formula (Hill Notation):
C15H11ClO7
CAS Number:
Molecular Weight:
338.70

Products

Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing