
Cyanidin 3-O-glucoside chloride

Synonym(s):
Kuromanin chloride, Asterin, Chrysanthemin, Cyanidin 3-O-glucoside chloride, Glucocyanidin chloride
Empirical Formula (Hill Notation):
C21H21ClO11
CAS Number:
Molecular Weight:
484.84
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing