
Chloroquine sulfate

Synonym(s):
Chloroquine sulfate monohydrate
Empirical Formula (Hill Notation):
C18H28ClN3O4S · H2O
CAS Number:
Molecular Weight:
435.97
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing