
CdSe/ZnS core-shell type quantum dots

Synonym(s):
Artificial atoms, Fluorescent nanocrystals, QDs
Linear Formula:
CdSe/ZnS
Products
Product Category
Brand
Physical Form
Quality Segment
Functional Group
Showing 1-20 of 45
Page 1 of 3
Compare
Product Number
Description
Pricing
Page 1 of 3