
Catharanthine

Synonym(s):
(+)-3,4-Didehydrocoronaridine, (2α,5β,6α,18β)-3,4-Didehydroibogamine-18-carboxylic acid methyl ester, 7-Ethyl-9,10,12,13-tetrahydro-6,9-methano-5H-pyrido[1′,2′:1,2]azepino[4,5-b]indole-6(6aH)-carboxylic acid methyl ester
Empirical Formula (Hill Notation):
C21H24N2O2
CAS Number:
Molecular Weight:
336.43
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing