
Carboxymethylcellulose sodium

Synonym(s):
Carboxymethylcellulose sodium salt, Sodium carboxymethylcellulose
CAS Number:
Products
Product Category
Brand
Color
Feature
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing