
Calcium lactobionate

Synonym(s):
Calcium lactobionate monohydrate, Lactobionic acid monohydrate
Empirical Formula (Hill Notation):
C24H42CaO24 · 2H2O
CAS Number:
Molecular Weight:
790.68
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing